Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614183 MIM: 610716 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DIS3L - DIS3 like exosome 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIPIN - TIMELESS interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289986.1 | 1048 | Intron | NP_001276915.1 | |||
NM_017858.2 | 1048 | Intron | NP_060328.2 | |||
XM_005254521.2 | 1048 | Missense Mutation | CCG,CTG | P,L 260 | XP_005254578.1 | |
XM_005254522.4 | 1048 | Missense Mutation | CCG,CTG | P,L 260 | XP_005254579.1 | |
XM_006720594.3 | 1048 | Missense Mutation | CCG,CTG | P,L 260 | XP_006720657.1 | |
XM_011521743.1 | 1048 | Missense Mutation | CCG,CTG | P,L 260 | XP_011520045.1 | |
XM_017022390.1 | 1048 | Missense Mutation | CCG,CTG | P,L 260 | XP_016877879.1 | |
XM_017022391.1 | 1048 | Intron | XP_016877880.1 |
Set Membership: |
HapMap |