Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603444 MIM: 613121 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FUBP1 - far upstream element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303433.1 | Intron | NP_001290362.1 | ||||
NM_003902.4 | Intron | NP_003893.2 | ||||
XM_011542391.2 | Intron | XP_011540693.1 | ||||
XM_011542392.2 | Intron | XP_011540694.1 | ||||
XM_011542393.2 | Intron | XP_011540695.1 | ||||
XM_017002739.1 | Intron | XP_016858228.1 | ||||
XM_017002740.1 | Intron | XP_016858229.1 | ||||
XM_017002741.1 | Intron | XP_016858230.1 | ||||
XM_017002742.1 | Intron | XP_016858231.1 | ||||
XM_017002743.1 | Intron | XP_016858232.1 | ||||
XM_017002744.1 | Intron | XP_016858233.1 |
NEXN - nexilin F-actin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |