Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613272 | ||||||||||||||||||||
Literature Links: |
KCTD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD3 - potassium channel tetramerization domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319294.1 | 982 | Missense Mutation | CGT,GGT | R,G 137 | NP_001306223.1 | |
NM_001319295.1 | 982 | Missense Mutation | CGT,GGT | R,G 35 | NP_001306224.1 | |
NM_016121.4 | 982 | Missense Mutation | CGT,GGT | R,G 137 | NP_057205.2 | |
XM_005273158.1 | 982 | Missense Mutation | CGT,GGT | R,G 35 | XP_005273215.1 |
Set Membership: |
JSNP |