Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600418 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMPH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AMPH - amphiphysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001635.3 | Intron | NP_001626.1 | ||||
NM_139316.2 | Intron | NP_647477.1 | ||||
XM_006715689.3 | Intron | XP_006715752.1 | ||||
XM_006715690.3 | Intron | XP_006715753.1 | ||||
XM_006715691.3 | Intron | XP_006715754.1 | ||||
XM_011515271.2 | Intron | XP_011513573.1 | ||||
XM_017011995.1 | Intron | XP_016867484.1 | ||||
XM_017011996.1 | Intron | XP_016867485.1 |
TRG-AS1 - T cell receptor gamma locus antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |