Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 608974 MIM: 611866 | |||||||||||||||||||||||||||||
Literature Links: |
C1RL PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
C1RL - complement C1r subcomponent like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297640.1 | Intron | NP_001284569.1 | ||||
NM_001297642.1 | Intron | NP_001284571.1 | ||||
NM_001297643.1 | Intron | NP_001284572.1 | ||||
NM_016546.3 | Intron | NP_057630.2 | ||||
XM_017019397.1 | Intron | XP_016874886.1 | ||||
XM_017019398.1 | Intron | XP_016874887.1 | ||||
XM_017019399.1 | Intron | XP_016874888.1 | ||||
XM_017019400.1 | Intron | XP_016874889.1 | ||||
XM_017019401.1 | Intron | XP_016874890.1 | ||||
XM_017019402.1 | Intron | XP_016874891.1 | ||||
XM_017019403.1 | Intron | XP_016874892.1 | ||||
XM_017019404.1 | Intron | XP_016874893.1 |
C1RL-AS1 - C1RL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBP5 - retinol binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |