Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601009 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TJP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TJP1 - tight junction protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301025.1 | Intron | NP_001287954.1 | ||||
NM_001301026.1 | Intron | NP_001287955.1 | ||||
NM_003257.4 | Intron | NP_003248.3 | ||||
NM_175610.3 | Intron | NP_783297.2 | ||||
XM_005254617.3 | Intron | XP_005254674.2 | ||||
XM_005254618.3 | Intron | XP_005254675.1 | ||||
XM_005254619.3 | Intron | XP_005254676.2 | ||||
XM_005254620.3 | Intron | XP_005254677.2 | ||||
XM_011521972.2 | Intron | XP_011520274.2 | ||||
XM_017022521.1 | Intron | XP_016878010.1 | ||||
XM_017022522.1 | Intron | XP_016878011.1 | ||||
XM_017022523.1 | Intron | XP_016878012.1 | ||||
XM_017022524.1 | Intron | XP_016878013.1 | ||||
XM_017022525.1 | Intron | XP_016878014.1 | ||||
XM_017022526.1 | Intron | XP_016878015.1 | ||||
XM_017022527.1 | Intron | XP_016878016.1 | ||||
XM_017022528.1 | Intron | XP_016878017.1 | ||||
XM_017022529.1 | Intron | XP_016878018.1 | ||||
XM_017022530.1 | Intron | XP_016878019.1 | ||||
XM_017022531.1 | Intron | XP_016878020.1 |
Set Membership: |
HapMap |