Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605999 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC10A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLEC10A - C-type lectin domain family 10 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256412.1 | 1597 | UTR 3 | XP_005256469.1 | |||
XM_011523613.1 | 1597 | UTR 3 | XP_011521915.1 | |||
XM_011523614.2 | 1597 | UTR 3 | XP_011521916.1 | |||
XM_011523615.1 | 1597 | UTR 3 | XP_011521917.1 |
Set Membership: |
HapMap |