Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3IL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAB3IL1 - RAB3A interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271686.1 | 615 | Missense Mutation | CAT,TAT | H,Y 297 | NP_001258615.1 | |
NM_013401.3 | 615 | Missense Mutation | CAT,TAT | H,Y 323 | NP_037533.2 | |
XM_005274140.4 | 615 | Missense Mutation | CAT,TAT | H,Y 370 | XP_005274197.1 | |
XM_006718647.3 | 615 | Missense Mutation | CAT,TAT | H,Y 370 | XP_006718710.1 | |
XM_011545196.2 | 615 | Missense Mutation | CAT,TAT | H,Y 370 | XP_011543498.1 | |
XM_011545197.2 | 615 | Missense Mutation | CAT,TAT | H,Y 329 | XP_011543499.1 | |
XM_011545198.2 | 615 | Missense Mutation | CAT,TAT | H,Y 323 | XP_011543500.1 | |
XM_011545199.2 | 615 | Missense Mutation | CAT,TAT | H,Y 297 | XP_011543501.1 | |
XM_011545200.2 | 615 | Missense Mutation | CAT,TAT | H,Y 175 | XP_011543502.1 | |
XM_011545201.2 | 615 | Intron | XP_011543503.1 |
Set Membership: |
HapMap |