Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146680 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDE PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IDE - insulin degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165946.1 | Intron | NP_001159418.1 | ||||
NM_001322793.1 | Intron | NP_001309722.1 | ||||
NM_001322794.1 | Intron | NP_001309723.1 | ||||
NM_001322795.1 | Intron | NP_001309724.1 | ||||
NM_001322796.1 | Intron | NP_001309725.1 | ||||
NM_001322797.1 | Intron | NP_001309726.1 | ||||
NM_004969.3 | Intron | NP_004960.2 | ||||
XM_017016187.1 | Intron | XP_016871676.1 | ||||
XM_017016188.1 | Intron | XP_016871677.1 | ||||
XM_017016189.1 | Intron | XP_016871678.1 | ||||
XM_017016190.1 | Intron | XP_016871679.1 |
Set Membership: |
HapMap |