Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 613081 MIM: 187680 | ||||||||||||||||||||||||||
Literature Links: |
KDM1B PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
KDM1B - lysine demethylase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153042.3 | Intron | NP_694587.3 | ||||
XM_005248925.1 | Intron | XP_005248982.1 | ||||
XM_005248926.1 | Intron | XP_005248983.1 | ||||
XM_005248928.1 | Intron | XP_005248985.1 | ||||
XM_011514385.1 | Intron | XP_011512687.1 | ||||
XM_011514386.1 | Intron | XP_011512688.1 | ||||
XM_017010440.1 | Intron | XP_016865929.1 | ||||
XM_017010441.1 | Intron | XP_016865930.1 | ||||
XM_017010442.1 | Intron | XP_016865931.1 | ||||
XM_017010443.1 | Intron | XP_016865932.1 | ||||
XM_017010445.1 | Intron | XP_016865934.1 | ||||
XM_017010446.1 | Intron | XP_016865935.1 |
TPMT - thiopurine S-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000367.3 | Intron | NP_000358.1 | ||||
XM_011514839.2 | Intron | XP_011513141.1 | ||||
XM_017011241.1 | Intron | XP_016866730.1 |
Set Membership: |
HapMap |