Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603717 MIM: 604013 MIM: 603456 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V0B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP6V0B - ATPase H+ transporting V0 subunit b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
B4GALT2 - beta-1,4-galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005417.2 | 60 | Intron | NP_001005417.1 | |||
NM_003780.4 | 60 | Intron | NP_003771.1 | |||
NM_030587.2 | 60 | UTR 5 | NP_085076.2 | |||
XM_017002716.1 | 60 | Intron | XP_016858205.1 | |||
XM_017002717.1 | 60 | Intron | XP_016858206.1 |
DPH2 - DPH2 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |