Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124075 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4B1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CYP4B1 - cytochrome P450 family 4 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000779.3 | Intron | NP_000770.2 | ||||
NM_001099772.1 | Intron | NP_001093242.1 | ||||
NM_001319161.1 | Intron | NP_001306090.1 | ||||
NM_001319162.1 | Intron | NP_001306091.1 | ||||
NM_001319163.1 | Intron | NP_001306092.1 | ||||
XM_011540832.1 | Intron | XP_011539134.1 | ||||
XM_011540833.1 | Intron | XP_011539135.1 | ||||
XM_017000466.1 | Intron | XP_016855955.1 |
Set Membership: |
HapMap JSNP |