Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPHP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPHP1 - nephrocystin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000272.3 | Intron | NP_000263.2 | ||||
NM_001128178.1 | Intron | NP_001121650.1 | ||||
NM_001128179.1 | Intron | NP_001121651.1 | ||||
NM_207181.2 | Intron | NP_997064.2 | ||||
XM_005263676.1 | Intron | XP_005263733.1 | ||||
XM_005263677.1 | Intron | XP_005263734.1 | ||||
XM_005263678.2 | Intron | XP_005263735.1 | ||||
XM_005263679.1 | Intron | XP_005263736.1 | ||||
XM_006712551.1 | Intron | XP_006712614.1 | ||||
XM_006712552.2 | Intron | XP_006712615.1 | ||||
XM_011511244.1 | Intron | XP_011509546.1 | ||||
XM_017004218.1 | Intron | XP_016859707.1 |