Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTCATCCTGCCCTGAACAAATC[A/G]GCTCAAGCCAGTCCCCCGCCAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602109 MIM: 613602 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928222 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928222 - uncharacterized LOC101928222 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MATN3 - matrilin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002381.4 | Intron | NP_002372.1 |
WDR35 - WD repeat domain 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |