Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600427 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
E2F3 - E2F transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243076.2 | Intron | NP_001230005.1 | ||||
NM_001949.4 | Intron | NP_001940.1 | ||||
XM_005248865.4 | Intron | XP_005248922.1 | ||||
XM_005248866.4 | Intron | XP_005248923.1 | ||||
XM_011514324.2 | Intron | XP_011512626.1 | ||||
XM_011514328.2 | Intron | XP_011512630.1 | ||||
XM_017010330.1 | Intron | XP_016865819.1 |
Set Membership: |
HapMap |