Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604209 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPNE5 - copine 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314017.1 | 2335 | Intron | NP_001300946.1 | |||
NM_001314018.1 | 2335 | UTR 3 | NP_001300947.1 | |||
NM_001314019.1 | 2335 | UTR 3 | NP_001300948.1 | |||
NM_001314020.1 | 2335 | UTR 3 | NP_001300949.1 | |||
NM_020939.1 | 2335 | UTR 3 | NP_065990.1 | |||
XM_005249247.2 | 2335 | UTR 3 | XP_005249304.1 | |||
XM_011514768.1 | 2335 | Intron | XP_011513070.1 | |||
XM_011514769.1 | 2335 | Intron | XP_011513071.1 | |||
XM_011514770.1 | 2335 | Intron | XP_011513072.1 | |||
XM_011514771.2 | 2335 | Intron | XP_011513073.1 | |||
XM_011514772.1 | 2335 | Intron | XP_011513074.1 | |||
XM_011514773.2 | 2335 | Intron | XP_011513075.1 | |||
XM_011514775.2 | 2335 | Intron | XP_011513077.1 | |||
XM_017011139.1 | 2335 | Intron | XP_016866628.1 |
RAB44 - RAB44, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |