Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCACCAGCTCTACTTCTTTACC[A/G]CCCTTTTAAAAGCCCTGATCTCTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608335 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371433 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105371433 - uncharacterized LOC105371433 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHO1 - pleckstrin homology domain containing O1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304722.1 | Intron | NP_001291651.1 | ||||
NM_001304723.1 | Intron | NP_001291652.1 | ||||
NM_001304724.1 | Intron | NP_001291653.1 | ||||
NM_016274.5 | Intron | NP_057358.2 | ||||
XM_017001420.1 | Intron | XP_016856909.1 |