Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153330 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRTP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRTP1 - growth hormone regulated TBC protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286732.1 | 2442 | Intron | NP_001273661.1 | |||
NM_001286733.1 | 2442 | UTR 3 | NP_001273662.1 | |||
NM_024719.3 | 2442 | UTR 3 | NP_078995.2 | |||
XM_005268333.3 | 2442 | UTR 3 | XP_005268390.1 | |||
XM_011537521.2 | 2442 | Intron | XP_011535823.1 | |||
XM_011537522.2 | 2442 | Intron | XP_011535824.1 | |||
XM_017020755.1 | 2442 | UTR 3 | XP_016876244.1 | |||
XM_017020756.1 | 2442 | UTR 3 | XP_016876245.1 |
LAMP1 - lysosomal associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005561.3 | 2442 | Intron | NP_005552.3 | |||
XM_011537494.1 | 2442 | Intron | XP_011535796.1 |
Set Membership: |
HapMap |