Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611782 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSCAML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSCAML1 - DS cell adhesion molecule like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020693.3 | 4626 | UTR 3 | NP_065744.2 | |||
XM_011542917.1 | 4626 | UTR 3 | XP_011541219.1 | |||
XM_011542918.1 | 4626 | UTR 3 | XP_011541220.1 | |||
XM_011542919.2 | 4626 | UTR 3 | XP_011541221.1 | |||
XM_011542920.2 | 4626 | UTR 3 | XP_011541222.1 | |||
XM_011542921.2 | 4626 | UTR 3 | XP_011541223.1 | |||
XM_011542924.2 | 4626 | UTR 3 | XP_011541226.1 | |||
XM_011542925.2 | 4626 | UTR 3 | XP_011541227.1 |