Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610264 MIM: 613276 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEPP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TEPP - testis, prostate and placenta expressed | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USB1 - U6 snRNA biogenesis phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF319 - zinc finger protein 319 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020807.2 | 2839 | UTR 3 | NP_065858.1 | |||
XM_005256069.3 | 2839 | UTR 3 | XP_005256126.1 |
Set Membership: |
HapMap |