Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTCCATTTAGCGGCTGGTTCA[C/T]AGATGGGCGGGGAAGAGCCGGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287944 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100287944 - uncharacterized LOC100287944 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIC8B - RIC8 guanine nucleotide exchange factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268998.3 | 17 | Intron | XP_005269055.1 | |||
XM_011538526.2 | 17 | Intron | XP_011536828.1 | |||
XM_011538527.2 | 17 | UTR 5 | XP_011536829.1 | |||
XM_011538528.2 | 17 | Intron | XP_011536830.1 | |||
XM_011538529.2 | 17 | Intron | XP_011536831.1 | |||
XM_017019548.1 | 17 | Intron | XP_016875037.1 | |||
XM_017019549.1 | 17 | Intron | XP_016875038.1 |