Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609011 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VASH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VASH1 - vasohibin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014909.4 | Intron | NP_055724.1 | ||||
XM_017021087.1 | Intron | XP_016876576.1 | ||||
XM_017021088.1 | Intron | XP_016876577.1 | ||||
XM_017021089.1 | Intron | XP_016876578.1 | ||||
XM_017021090.1 | Intron | XP_016876579.1 | ||||
XM_017021091.1 | Intron | XP_016876580.1 |