Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604282 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
H1FOO PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
H1FOO - H1 histone family member O, oocyte specific | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLXND1 - plexin D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015103.2 | Intron | NP_055918.2 | ||||
XM_011512588.1 | Intron | XP_011510890.1 | ||||
XM_011512589.1 | Intron | XP_011510891.1 | ||||
XM_011512590.2 | Intron | XP_011510892.1 | ||||
XM_011512592.1 | Intron | XP_011510894.1 |