Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612962 MIM: 610355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCTN5 - dynactin subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199011.1 | Intron | NP_001185940.1 | ||||
NM_001199743.1 | Intron | NP_001186672.1 | ||||
NM_032486.3 | Intron | NP_115875.1 |
PALB2 - partner and localizer of BRCA2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024675.3 | Intron | NP_078951.2 | ||||
XM_011545946.2 | Intron | XP_011544248.1 | ||||
XM_011545947.2 | Intron | XP_011544249.1 | ||||
XM_011545948.2 | Intron | XP_011544250.1 | ||||
XM_017023671.1 | Intron | XP_016879160.1 | ||||
XM_017023672.1 | Intron | XP_016879161.1 | ||||
XM_017023673.1 | Intron | XP_016879162.1 |