Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCGCAGCTCCCGCTTCTCCCTG[T/C]GCTCCTCCATCTCCTCGATGTCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605367 | ||||||||||||||||||||
Literature Links: |
ARHGAP44 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP44 - Rho GTPase activating protein 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELAC2 - elaC ribonuclease Z 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165962.1 | 2456 | Missense Mutation | CAC,CGC | H,R 741 | NP_001159434.1 | |
NM_018127.6 | 2456 | Missense Mutation | CAC,CGC | H,R 781 | NP_060597.4 | |
NM_173717.1 | 2456 | Missense Mutation | CAC,CGC | H,R 780 | NP_776065.1 |