Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTATCTCTTCTCCCTGCCCAT[C/T]GAGGAGTGTGATATCATTAGCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS2P45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS2P45 - ribosomal protein S2 pseudogene 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TANGO6 - transport and golgi organization 6 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024562.1 | Intron | NP_078838.1 | ||||
XM_011523327.2 | Intron | XP_011521629.1 |