Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAAGGAAGACACAGAAACGAAG[A/C]AAGAGCATGCTTCCTGGAGTTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLE1 - notchless homolog 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC45B - unc-45 myosin chaperone B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033576.1 | Intron | NP_001028748.1 | ||||
NM_001267052.1 | Intron | NP_001253981.1 | ||||
NM_001308281.1 | Intron | NP_001295210.1 | ||||
NM_173167.3 | Intron | NP_775259.1 | ||||
XM_017024234.1 | Intron | XP_016879723.1 |