Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTAATATCTCAGCATCGCGAACC[A/G]GGCCTCGGATTTGCTGCGCAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM96A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM96A - family with sequence similarity 96 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX1 - sorting nexin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242933.1 | Intron | NP_001229862.1 | ||||
NM_003099.4 | Intron | NP_003090.2 | ||||
NM_148955.3 | Intron | NP_683758.1 | ||||
XM_005254611.2 | Intron | XP_005254668.1 |