Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCTCTCTTCTCTTTCTCTCTC[C/T]GGAAACGTGAGTAGCTAATCGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616759 MIM: 604429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOSIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOSIP - nitric oxide synthase interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270960.1 | 83 | Intron | NP_001257889.1 | |||
NM_015953.4 | 83 | Intron | NP_057037.1 | |||
XM_005258964.4 | 83 | Intron | XP_005259021.1 | |||
XM_006723235.3 | 83 | Intron | XP_006723298.1 | |||
XM_011527015.2 | 83 | Intron | XP_011525317.1 | |||
XM_011527017.2 | 83 | Intron | XP_011525319.1 | |||
XM_017026851.1 | 83 | Intron | XP_016882340.1 | |||
XM_017026852.1 | 83 | Intron | XP_016882341.1 |
PRRG2 - proline rich and Gla domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000951.2 | 83 | Intron | NP_000942.1 | |||
NM_001316335.1 | 83 | Intron | NP_001303264.1 | |||
XM_006723286.2 | 83 | UTR 5 | XP_006723349.1 | |||
XM_011527124.1 | 83 | Intron | XP_011525426.1 |