Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCGCCTGCCCCGCGCGCCCTGC[C/G]GGAGGTCGGCGCTGAGCTTGCGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154582 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN2A1 - mannosidase alpha class 2A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002372.3 | 189 | UTR 5 | NP_002363.2 | |||
XM_011543395.2 | 189 | UTR 5 | XP_011541697.1 | |||
XM_017009472.1 | 189 | Intron | XP_016864961.1 |