Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCTCAGGAGGCCACGCTGCTCC[C/T]CGGCGGAAACCAAAGTGCCTTCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616574 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378614 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105378614 - uncharacterized LOC105378614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MINOS1 - mitochondrial inner membrane organizing system 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032363.3 | Intron | NP_001027535.1 | ||||
NM_001204082.1 | Intron | NP_001191011.1 | ||||
NM_001204083.1 | Intron | NP_001191012.1 |
MINOS1-NBL1 - MINOS1-NBL1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204088.1 | Intron | NP_001191017.1 | ||||
NM_001204089.1 | Intron | NP_001191018.1 |