Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCATTCATATTTTCAGTATAGC[C/T]TATCACATAATTGCCTTTTGGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600563 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGFR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTGFR - prostaglandin F receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |