Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAACCAGGGAAAGAACGGATTCT[C/T]ACCATAGATACCATTTTTGAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612842 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASD2 - RASD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014310.3 | 2397 | Intron | NP_055125.2 | |||
XM_005261442.4 | 2397 | Intron | XP_005261499.1 | |||
XM_011530040.2 | 2397 | Intron | XP_011528342.1 | |||
XM_017028701.1 | 2397 | UTR 5 | XP_016884190.1 | |||
XM_017028702.1 | 2397 | Intron | XP_016884191.1 |