Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCCCTGGGACGCGGCGCTCTCC[C/T]GGCGCTGCTGCCTCGGCTTGGTCTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600600 | |||||||||||||||||||||||
Literature Links: |
EPHB1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EPHB1 - EPH receptor B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004441.4 | 327 | UTR 5 | NP_004432.1 | |||
XM_011512542.1 | 327 | Intron | XP_011510844.1 | |||
XM_017005866.1 | 327 | UTR 5 | XP_016861355.1 | |||
XM_017005867.1 | 327 | Intron | XP_016861356.1 | |||
XM_017005868.1 | 327 | Intron | XP_016861357.1 |