Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCAGCAGCTGCCCGTCTTCCC[A/G]CATAGAGGGCAGGAAGGACAACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAD1 - adenosine deaminase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159285.1 | 219 | Intron | NP_001152757.1 | |||
NM_001159295.1 | 219 | UTR 5 | NP_001152767.1 | |||
NM_139243.3 | 219 | Intron | NP_640336.1 | |||
XM_005262741.2 | 219 | UTR 5 | XP_005262798.1 | |||
XM_005262743.2 | 219 | Intron | XP_005262800.1 | |||
XM_005262744.2 | 219 | UTR 5 | XP_005262801.1 | |||
XM_005262745.2 | 219 | UTR 5 | XP_005262802.1 | |||
XM_011531603.1 | 219 | Intron | XP_011529905.1 |