Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGCGGAGAGAAGCCCCGGAGA[A/G]GACAAGGTGAAGAGCCAATGGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600659 MIM: 614117 MIM: 605235 | ||||||||||||||||||||
Literature Links: |
E2F4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
E2F4 - E2F transcription factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EXOC3L1 - exocyst complex component 3 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178516.3 | 320 | Intron | NP_848611.2 |
KIAA0895L - KIAA0895 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040715.1 | 320 | UTR 5 | NP_001035805.1 |
NOL3 - nucleolar protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |