Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAAACCCAGGCCGCAGCGCCC[C/T]GGCCCGACTGGTCCTGTCCTTCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF15 - Rho guanine nucleotide exchange factor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025014.1 | Intron | NP_079290.1 | ||||
NM_173728.3 | Intron | NP_776089.2 | ||||
XM_011523734.1 | Intron | XP_011522036.1 | ||||
XM_011523735.1 | Intron | XP_011522037.1 | ||||
XM_011523736.1 | Intron | XP_011522038.1 | ||||
XM_011523737.1 | Intron | XP_011522039.1 |