Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCAGGAGCTGGCCAGCTGATGG[C/T]CTCCATTTTTTCCTACAGGATTTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604544 | |||||||||||||||||||||||
Literature Links: |
LARS2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LARS2 - leucyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015340.3 | Intron | NP_056155.1 | ||||
XM_005265006.1 | Intron | XP_005265063.1 | ||||
XM_011533554.2 | Intron | XP_011531856.1 | ||||
XM_017006042.1 | Intron | XP_016861531.1 |