Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCCAGCCCGCCCGTCGCCCGC[C/T]CCGGGAAAGGACCCGCTATCGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604186 MIM: 616150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCL14 - C-X-C motif chemokine ligand 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004887.4 | 139 | UTR 5 | NP_004878.2 |
SLC25A48 - solute carrier family 25 member 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145282.4 | 139 | Intron | NP_660325.4 | |||
XM_005271903.1 | 139 | Intron | XP_005271960.1 | |||
XM_006714544.1 | 139 | Intron | XP_006714607.1 | |||
XM_006714546.1 | 139 | Intron | XP_006714609.1 | |||
XM_006714550.1 | 139 | Intron | XP_006714613.1 | |||
XM_006714552.2 | 139 | Intron | XP_006714615.1 | |||
XM_011543196.1 | 139 | Intron | XP_011541498.1 | |||
XM_011543199.1 | 139 | Intron | XP_011541501.1 | |||
XM_017009086.1 | 139 | Intron | XP_016864575.1 | |||
XM_017009087.1 | 139 | Intron | XP_016864576.1 | |||
XM_017009088.1 | 139 | Intron | XP_016864577.1 | |||
XM_017009089.1 | 139 | Intron | XP_016864578.1 | |||
XM_017009090.1 | 139 | UTR 5 | XP_016864579.1 | |||
XM_017009091.1 | 139 | Intron | XP_016864580.1 | |||
XM_017009092.1 | 139 | UTR 5 | XP_016864581.1 |