Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGGTTATCAGAAATTTAGAACC[A/G]CTCCACTTGGGAGACAAAGGGCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRIP3 - cysteine rich protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_206922.2 | 1757 | Intron | NP_996805.2 | |||
XM_005249103.3 | 1757 | UTR 5 | XP_005249160.1 | |||
XM_011514609.2 | 1757 | UTR 5 | XP_011512911.1 | |||
XM_017010856.1 | 1757 | UTR 5 | XP_016866345.1 | |||
XM_017010857.1 | 1757 | UTR 5 | XP_016866346.1 |
SLC22A7 - solute carrier family 22 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |