Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCGCGAGATCGGATCTGGGCTC[C/T]GCGAGGTTTTGGCGTAGTTGTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603631 MIM: 608754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS9 - ribosomal protein S9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013.3 | Intron | NP_001004.2 | ||||
NM_001321701.1 | Intron | NP_001308630.1 | ||||
NM_001321702.1 | Intron | NP_001308631.1 | ||||
NM_001321704.1 | Intron | NP_001308633.1 | ||||
NM_001321705.1 | Intron | NP_001308634.1 | ||||
NM_001321706.1 | Intron | NP_001308635.1 | ||||
XM_017027112.1 | Intron | XP_016882601.1 |
TSEN34 - tRNA splicing endonuclease subunit 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |