Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCACCGGCGGTGCACTTTCTGT[C/G]GATACGGTAGCTTCGCGCTTGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608451 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETHE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETHE1 - ETHE1, persulfide dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF575 - zinc finger protein 575 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174945.2 | 10 | Intron | NP_777605.1 | |||
XM_005258783.3 | 10 | UTR 5 | XP_005258840.1 | |||
XM_011526793.2 | 10 | Intron | XP_011525095.1 |