Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTTTAAAAATAATAAAACTTCA[A/G]AAAGAACAAAAACAACCCCAAACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600483 MIM: 604039 MIM: 606456 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF8 - fibroblast growth factor 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGEA5 - meningioma expressed antigen 5 (hyaluronidase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142434.1 | 4409 | UTR 3 | NP_001135906.1 | |||
NM_012215.3 | 4409 | UTR 3 | NP_036347.1 | |||
XM_017015585.1 | 4409 | UTR 3 | XP_016871074.1 | |||
XM_017015586.1 | 4409 | UTR 3 | XP_016871075.1 |
NPM3 - nucleophosmin/nucleoplasmin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |