Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTAGAAATACATTGTTACATT[A/T]AAAAATGCTGTTATTTGAAAGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300806 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFF2 - AF4/FMR2 family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169122.1 | Intron | NP_001162593.1 | ||||
NM_001169123.1 | Intron | NP_001162594.1 | ||||
NM_001169124.1 | Intron | NP_001162595.1 | ||||
NM_001169125.1 | Intron | NP_001162596.1 | ||||
NM_001170628.1 | Intron | NP_001164099.1 | ||||
NM_002025.3 | Intron | NP_002016.2 |