Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTGAGGGAATTAAATTCTTTG[A/C]ACGGATGCGCTGGTTTTCGGAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLN - neurolysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020726.4 | Intron | NP_065777.1 | ||||
XM_005248559.2 | Intron | XP_005248616.1 | ||||
XM_006714661.2 | Intron | XP_006714724.1 | ||||
XM_017009673.1 | Intron | XP_016865162.1 |
SGTB - small glutamine rich tetratricopeptide repeat containing beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019072.2 | Intron | NP_061945.1 | ||||
XM_005248548.3 | Intron | XP_005248605.1 | ||||
XM_017009597.1 | Intron | XP_016865086.1 |