Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAGAGGACAGAGCTGGGTAGTA[C/T]GCCTCTGCTGGGCGCGGCCTGGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNBIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTNNBIP1 - catenin beta interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012329.1 | 1780 | UTR 3 | NP_001012329.1 | |||
NM_020248.2 | 1780 | UTR 3 | NP_064633.1 | |||
XM_017001853.1 | 1780 | UTR 3 | XP_016857342.1 | |||
XM_017001854.1 | 1780 | UTR 3 | XP_016857343.1 | |||
XM_017001855.1 | 1780 | UTR 3 | XP_016857344.1 |