Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGTTTATCTAAGTAGTTTTAAG[A/G]AAAAGAGGAGCCTATGGTTTGCTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182389 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724058 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102724058 - uncharacterized LOC102724058 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN1A - sodium voltage-gated channel alpha subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165963.1 | 8196 | UTR 3 | NP_001159435.1 | |||
NM_001165964.1 | 8196 | UTR 3 | NP_001159436.1 | |||
NM_001202435.1 | 8196 | UTR 3 | NP_001189364.1 | |||
NM_006920.4 | 8196 | UTR 3 | NP_008851.3 | |||
XM_011511602.2 | 8196 | UTR 3 | XP_011509904.1 | |||
XM_011511604.2 | 8196 | Intron | XP_011509906.1 | |||
XM_011511605.2 | 8196 | UTR 3 | XP_011509907.1 | |||
XM_011511606.2 | 8196 | UTR 3 | XP_011509908.1 | |||
XM_017004644.1 | 8196 | UTR 3 | XP_016860133.1 | |||
XM_017004645.1 | 8196 | Intron | XP_016860134.1 | |||
XM_017004646.1 | 8196 | Intron | XP_016860135.1 | |||
XM_017004647.1 | 8196 | Intron | XP_016860136.1 | |||
XM_017004648.1 | 8196 | Intron | XP_016860137.1 | |||
XM_017004649.1 | 8196 | Intron | XP_016860138.1 | |||
XM_017004650.1 | 8196 | UTR 3 | XP_016860139.1 | |||
XM_017004651.1 | 8196 | UTR 3 | XP_016860140.1 | |||
XM_017004652.1 | 8196 | UTR 3 | XP_016860141.1 | |||
XM_017004653.1 | 8196 | UTR 3 | XP_016860142.1 | |||
XM_017004654.1 | 8196 | UTR 3 | XP_016860143.1 |
Set Membership: |
HapMap |