Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTGTAAAGGGCTGGGTGCGAC[A/G]GGGCACCAGGCCATAGAATTCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF1 - C1q and tumor necrosis factor related protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030968.3 | Intron | NP_112230.1 | ||||
NM_153372.1 | Intron | NP_699203.1 | ||||
NM_198593.3 | Intron | NP_940995.1 | ||||
NM_198594.2 | Intron | NP_940996.1 | ||||
XM_006721663.1 | Intron | XP_006721726.1 | ||||
XM_006721664.1 | Intron | XP_006721727.1 | ||||
XM_006721666.1 | Intron | XP_006721729.1 | ||||
XM_017024126.1 | Intron | XP_016879615.1 |
C1QTNF1-AS1 - C1QTNF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |