Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCGCCCTATTCGCCATCTTAAC[A/G]TAGGGCATTCCCCACAGCTGCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613734 MIM: 612981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC115 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC115 - coiled-coil domain containing 115 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321118.1 | 574 | Intron | NP_001308047.1 | |||
NM_001321119.1 | 574 | Intron | NP_001308048.1 | |||
NM_032357.3 | 574 | Intron | NP_115733.2 |
IMP4 - IMP4 homolog, U3 small nucleolar ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320304.1 | 574 | UTR 5 | NP_001307233.1 | |||
NM_001320305.1 | 574 | UTR 5 | NP_001307234.1 | |||
NM_001320306.1 | 574 | UTR 5 | NP_001307235.1 | |||
NM_001320307.1 | 574 | UTR 5 | NP_001307236.1 | |||
NM_001320309.1 | 574 | UTR 5 | NP_001307238.1 | |||
NM_001320310.1 | 574 | UTR 5 | NP_001307239.1 | |||
NM_001320311.1 | 574 | UTR 5 | NP_001307240.1 | |||
NM_033416.2 | 574 | UTR 5 | NP_219484.1 |