Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACTTTGGGAGTTAAGTTCAGCA[C/T]TGGATTCCACCCCGCCACCCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HKR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HKR1 - HKR1, GLI-Kruppel zinc finger family member | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF527 - zinc finger protein 527 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032453.1 | Intron | NP_115829.1 | ||||
XM_005259328.4 | Intron | XP_005259385.1 | ||||
XM_005259329.3 | Intron | XP_005259386.1 | ||||
XM_017027380.1 | Intron | XP_016882869.1 | ||||
XM_017027381.1 | Intron | XP_016882870.1 |